missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMP19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19423-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| HMP19 | |
| Polyclonal | |
| Unconjugated | |
| NSG2 | |
| HMP19, HMP19 protein, NSG2, Protein p19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18197, 497878, 51617 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P47759, Q9Y328 | |
| NSG2 | |
| HMP19 Fusion Protein Ag13751 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction