missing translation for 'onlineSavingsMsg'
Learn More

HMN-176, MedChemExpress

Product Code. 16453376
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453376 25 mg 25mg
16451947 50 mg 50mg
16428467 5 mg 5mg
16425798 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16453376 Supplier Medchem Express Supplier No. HY13647
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress HMN-176 is a stilbene derivative which inhibits mitosis, interfering with polo-like kinase-1 (plk1), without significant effect on tubulin polymerization.

HMN-176 is a stilbene derivative which inhibits mitosis, interfering with polo-like kinase-1 (plk1), without significant effect on tubulin polymerization.

Specifications

Chemical Name or Material HMN-176
CAS 173529-10-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H18N2O4S
Quantity 25 mg
Solubility Information DMSO : ≥ 30 mg/mL (78.45 mM)
SMILES O=S(C1=CC=C(OC)C=C1)(NC2=CC=CC=C2/C=C/C3=CC=N(C=C3)=O)=O
Molecular Weight (g/mol) 382.43
Formula Weight 382.43
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.