missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMHA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14832-1-AP-20UL
This item is not returnable.
View return policy
Description
HMHA1 (ARHGAP45) Contains a GTPase activator for the Rho-type GTPases (RhoGAP) domain that would be able to negatively regulate the actin cytoskeleton as well as cell spreading. However, also contains N-terminally a BAR-domin which is able to play an autoinhibitory effect on this RhoGAP activity.Specifications
| HMHA1 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP45 | |
| HA 1, HLA HA1, HMHA1, KIAA0223, mHag HA 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23526, 314618, 70719 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3TBD2, Q92619 | |
| Arhgap45 | |
| HMHA1 Fusion Protein Ag6636 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction