Learn More
HMHA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14832-1-AP-150UL
 View more versions of this product
Product Code. 16873684
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
HMHA1 (ARHGAP45) Contains a GTPase activator for the Rho-type GTPases (RhoGAP) domain that would be able to negatively regulate the actin cytoskeleton as well as cell spreading. However, also contains N-terminally a BAR-domin which is able to play an autoinhibitory effect on this RhoGAP activity.
Specifications
Specifications
HMHA1
Polyclonal
Unconjugated
ARHGAP45
HA 1, HLA HA1, HMHA1, KIAA0223, mHag HA 1
Rabbit
Antigen Affinity Chromatography
RUO
23526, 314618, 70719
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3TBD2, Q92619
Arhgap45
HMHA1 Fusion Protein Ag6636
150 μL
Primary
Human, Mouse, Rat