Learn More
HMHA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544232
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MMHMQTAPLP VHFQMLCESS KLYDPGQQYA SHVRQLQRDQ EPDVHYDFEP Sequence homology: Cow: 100%; Dog: 100%; Horse: 75%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 75%; Rat: 100%; Zebrafish: 92%
HMHA1 Polyclonal antibody specifically detects HMHA1 in Human samples. It is validated for Western Blot.Specifications
HMHA1 | |
Polyclonal | |
Unconjugated | |
ARHGAP45 | |
6330406L22Rik; ARHGAP45; AW539505; Ha-1; histocompatibility (minor) HA-1; HLA-HA1; Hmha1; KIAA0223; mHag HA-1; Minor histocompatibility antigen HA-1; minor histocompatibility protein HA-1; minor histocompatibility protein HA-1; rho GTPase-activating protein 45; RGD1308662; Rho GTPase activating protein 45; rho GTPase-activating protein 45 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23526 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q92619 | |
ARHGAP45 | |
Synthetic peptide directed towards the N-terminal of human HMHA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |