Learn More
HMGXB4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA551448
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: YDDSVKKEDC FDGDHTFEDI GLAAGRSQRE KKRSYKDFLR EEEEIAAQVR NSSKKKLKDS ELYFLGTDTH KKKRKHSSDD YYYGDISSLE SSQKKKKKSS PQSTDTAMDL LKAITSPLAA GSKPSKKTGE KSSGSSSHSE S Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
High mobility group (HMG) proteins are nonhistone chromosomal proteins.Spécification
HMGXB4 | |
Polyclonal | |
Unconjugated | |
HMGXB4 | |
high mobility group protein 2-like 1; high-mobility group protein 2-like 1; HMG box domain containing 4; HMG box-containing protein 4; HMG domain-containing protein 4; HMG2L1; HMGBCG; HMG-box containing 4; HMGXB4; Protein HMGBCG; Protein THC211630; THC211630 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10042 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UGU5 | |
HMGXB4 | |
Recombinant protein corresponding to Human HMGXB4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only