missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGN3/TRIP7 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | HMGN3/TRIP7 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
HMGN3/TRIP7 Polyclonal specifically detects HMGN3/TRIP7 in Mouse samples. It is validated for Western Blot.Specifications
| HMGN3/TRIP7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| DKFZp686E20226, high mobility group nucleosomal binding domain 3, high mobility group nucleosome-binding domain-containing protein 3, high-mobility group protein HMGN3, PNAS-25, thyroid hormone receptor interacting protein 7, thyroid hormone receptor interactor 7, Thyroid receptor-interacting protein 7, TR-interacting protein 7, TRIP-7, TRIP7FLJ42187 | |
| The immunogen is a synthetic peptide directed towards the middle region of Mouse HMGN3/TRIP7 (NP_080398). Peptide sequence SKPRKTSAKKEPGTKISRGAKGKKEEKQEAGEEGTAPSANGDTKVEEAQR | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 9324 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title