missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGN3/TRIP7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62640
This item is not returnable.
View return policy
Description
HMGN3/TRIP7 Polyclonal antibody specifically detects HMGN3/TRIP7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| HMGN3/TRIP7 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZp686E20226, high mobility group nucleosomal binding domain 3, high mobility group nucleosome-binding domain-containing protein 3, high-mobility group protein HMGN3, PNAS-25, thyroid hormone receptor interacting protein 7, thyroid hormone receptor interactor 7, Thyroid receptor-interacting protein 7, TR-interacting protein 7, TRIP-7, TRIP7FLJ42187 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MPKRKSPENTEGKDGSKVTKQEPTRRSA | |
| 100 μg | |
| Cell Biology, Signal Transduction | |
| 9324 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction