missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551183
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total HMGN3 protein.
Binds to nucleosomes, regulating chromatin structure and consequently, chromatin-dependent processes such as transcription, DNA replication and DNA repair. Affects both insulin and glucagon levels and modulates the expression of pancreatic genes involved in insulin secretion. Regulates the expression of the glucose transporter SLC2A2 by binding specifically to its promoter region and recruiting PDX1 and additional transcription factors. Regulates the expression of SLC6A9, a glycine transporter which regulates the glycine concentration in synaptic junctions in the central nervous system, by binding to its transcription start site. May play a role in ocular development and astrocyte function.
Specifications
| HMGN3 | |
| Polyclonal | |
| Unconjugated | |
| HMGN3 | |
| 1110002A15Rik; 6330514M13Rik; BB071015; high mobility group nucleosomal binding domain 3; high mobility group nucleosome-binding domain-containing protein 3; HMGN3; PNAS-24; PNAS-25; thyroid hormone receptor interacting protein 7; Thyroid receptor-interacting protein 7; TR-interacting protein 7; TRIP7; TRIP-7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9324, 94353 | |
| -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q15651, Q9DCB1 | |
| HMGN3 | |
| Full length fusion protein of human HMGN3. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction