missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGN2 Recombinant Rabbit Monoclonal Antibody (JE53-85)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536161
This item is not returnable.
View return policy
Description
HMGN2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene binds nucleosomal DNA and is associated with transcriptionally active chromatin. Along with a similar protein, HMGN1, the encoded protein may help maintain an open chromatin configuration around transcribable genes. The protein has also been found to have antimicrobial activity against bacteria, viruses and fungi.
Specifications
| HMGN2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P05204 | |
| HMGN2 | |
| Recombinant protein within Human HMGN2 aa 1-90. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE53-85 | |
| Unconjugated | |
| HMGN2 | |
| high mobility group nucleosomal binding domain 2; high mobility group nucleosome binding domain 2; high mobility group nucleosome-binding domain-containing protein 2; high mobility group protein 17; high mobility group protein N2; high-mobility group (nonhistone chromosomal) protein 17; high-mobility group nucleosomal binding domain 2; high-mobility group nucleosome-binding domain-containing protein 2; HMG17; Hmg-17; Hmgn2; Non-histone chromosomal protein HMG-17; nonhistone chromosomal protein HMG-17 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3151 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction