missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585030
This item is not returnable.
View return policy
Description
Immunogen sequence: PKRKAKGDAK GDKAKVKDEP Q Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 90%.
The protein encoded by this gene binds nucleosomal DNA and is associated with transcriptionally active chromatin. Along with a similar protein, HMGN1, the encoded protein may help maintain an open chromatin configuration around transcribable genes. The protein has also been found to have antimicrobial activity against bacteria, viruses and fungi.
Specifications
| HMGN2 | |
| Polyclonal | |
| Unconjugated | |
| HMGN2 | |
| high mobility group nucleosomal binding domain 2; high mobility group nucleosome binding domain 2; high mobility group nucleosome-binding domain-containing protein 2; high mobility group protein 17; high mobility group protein N2; high-mobility group (nonhistone chromosomal) protein 17; high-mobility group nucleosomal binding domain 2; high-mobility group nucleosome-binding domain-containing protein 2; HMG17; Hmg-17; Hmgn2; Non-histone chromosomal protein HMG-17; nonhistone chromosomal protein HMG-17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3151 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P05204 | |
| HMGN2 | |
| Recombinant protein corresponding to Human HMGN2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction