missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGCS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543197
This item is not returnable.
View return policy
Description
Peptide sequence: KDFTLNDFGF MIFHSPYCKL VQKSLARMLL NDFLNDQNRD KNSIYSGLEA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
This enzyme condenses acetyl-CoA with acetoacetyl-CoA to form HMG-CoA, which is the substrate for HMG-CoA reductase.
Specifications
| HMGCS1 | |
| Polyclonal | |
| Unconjugated | |
| HMGCS1 | |
| 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 1; 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble) isoform 1; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble) isoform 2; B130032C06Rik; HMG-CoA synthase; HMGCS; Hmgcs1; HMGCS1 protein-like; hydroxymethylglutaryl-CoA synthase 1; hydroxymethylglutaryl-CoA synthase, cytoplasmic; Unknown (protein for IMAGE:7955111) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3157 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q01581 | |
| HMGCS1 | |
| Synthetic peptide directed towards the middle region of human HMGCS1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction