missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGCS1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-36554CL1
This item is not returnable.
View return policy
Description
HMGCS1 Polyclonal antibody specifically detects HMGCS1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| HMGCS1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase, 3-hydroxy-3-methylglutaryl coenzyme A synthase, 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble), 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble), EC 2.3.3.10, HMG-CoA synthase, HMGCS, hydroxymethylglutaryl-CoA synthase, cytoplasmic, MGC90332 | |
| The immunogen recognized by the antibody maps to a region between residue 470 to 520 of human 3-hydroxy-3-methylglutaryl-CoA Synthase 1 using the numbering given in entry NP_001091742.1. | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 3157 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction