missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGCL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16898-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the HMG-CoA lyase family. It is a mitochondrial enzyme that catalyzes the final step of leucine degradation and plays a key role in ketone body formation. Mutations in this gene are associated with HMG-CoA lyase deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| HMGCL | |
| Polyclonal | |
| Unconjugated | |
| HMGCL | |
| HL, HMG CoA lyase, HMGCL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15356, 3155, 79238 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35914, P38060, P97519 | |
| HMGCL | |
| HMGCL Fusion Protein Ag10426 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction