Learn More
Invitrogen™ HMGCL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596910
Description
Immunogen sequence: MAAMRKALPR RLVGLASLRA VSTSSMGTLP KRVKIVEVGP RDGLQNEKNI VSTPVKIKLI DMLSEAGLSV IETTSFVSPK WVPQMGDHTE VLKGIQKFPG INYPVLTPNL KGFEAAVAAG AKEVVIFGAA SELFTKKNIN CSIEESFQRF DAILKAAQSA NISVRGYVSC ALGCPYEGKI SPAKVAEVTK KFYSMGCYEI SLGDTIGVGT PGIMKDMLSA VMQEVPLAAL AVHCHDTYGQ ALANTLMALQ MGVSVVDSSV AGLGGCPYAQ GASGNLATED LVYMLEGLGI HTGVNLQKLL EAGNFICQAL NRKTSSKVAQ ATCKL; Positive Samples: U-251MG, SKOV3, BT-474, MCF-7, Mouse liver, Mouse kidney; Cellular Location: Mitochondrion matrix, Peroxisome.
The protein encoded by this gene belongs to the HMG-CoA lyase family. It is a mitochondrial enzyme that catalyzes the final step of leucine degradation and plays a key role in ketone body formation. Mutations in this gene are associated with HMG-CoA lyase deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| HMGCL | |
| Polyclonal | |
| Unconjugated | |
| HMGCL | |
| 3-hydroxy-3-methylglutarate-CoA lyase; 3-hydroxy-3-methylglutaryl-CoA lyase; 3-hydroxy-3-methylglutaryl-Coenzyme A lyase; 3-hydroxymethyl-3-methylglutaryl-CoA lyase; 3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase; AW476067; HL; Hmgcl; HMG-CoA lyase; hydroxymethylglutaricaciduria; hydroxymethylglutaryl-CoA lyase, mitochondrial; mitochondrial 3-hydroxy-3-methylglutaryl-CoA lyase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15356, 3155, 79238 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35914, P38060, P97519 | |
| HMGCL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-325 of human HMGCL (NP_0001822). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.