missing translation for 'onlineSavingsMsg'
Learn More

HMGB4 Antibody (OTI5G3) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30688652 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688652 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688652 Supplier Novus Biologicals Supplier No. NBP344707MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMGB4 Monoclonal antibody specifically detects HMGB4 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB4
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone OTI5G3
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias dJ1007G16.5, FLJ40388, high mobility group box 4, high mobility group protein B4, HMG2 like, MGC88128
Host Species Mouse
Immunogen Full length human recombinant protein of human HMGB4(NP_001008728) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127540
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.