missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584556
This item is not returnable.
View return policy
Description
Immunogen sequence: EKYEKDVADY KSKGKFDGAK GPAKVARKKV EEEDEE Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.
HMGB3 belongs to the high mobility group protein superfamily. Like HMG1 and HMG2, HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription.
Specifications
| HMGB3 | |
| Polyclonal | |
| Unconjugated | |
| HMGB3 | |
| high mobility group box 3; high mobility group protein 1; high mobility group protein 2a; High mobility group protein 4; high mobility group protein B1; high mobility group protein B3; high mobilty group protein 2A; high-mobility group (nonhistone chromosomal) protein 4; high-mobility group protein 4; HMG-1; HMG2a; HMG-2a; HMG4; HMG-4; HMGB1; Hmgb3; HMGB3 transcript variant 1/2; MGC90319; RGD1564407; Unknown (protein for MGC:133786) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3149 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15347 | |
| HMGB3 | |
| Recombinant protein corresponding to Human HMGB3. Recombinant protein control fragment (Product #RP-101986). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction