missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3/HMG4 Rabbit anti-Human, Mouse, Rat, Clone: S01-9C9, Novus Biologicals™
Shop All Bio Techne ProductsDescription
HMGB3/HMG4 Monoclonal antibody specifically detects HMGB3/HMG4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
Specifications
| Antigen | HMGB3/HMG4 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | S01-9C9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 20 μg/mL |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide |
| Gene Alias | High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human HMGB3/HMG4 (Uniprot#: O15347) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?