missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3/HMG4 Antibody (JE63-39), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32437
This item is not returnable.
View return policy
Description
HMGB3/HMG4 Monoclonal antibody specifically detects HMGB3/HMG4 in Human samples. It is validated for Western Blot
Specifications
| HMGB3/HMG4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein | |
| Synthetic peptide within Human HMG4 aa 50-150. (Uniprot: O15347) | |
| 100 μL | |
| Chromatin Research, DNA Repair | |
| 3149 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE63-39 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction