missing translation for 'onlineSavingsMsg'
Learn More

HMGB1/HMG-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18373485 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373485 100 μg 100µL
18312597 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18373485 Supplier Novus Biologicals Supplier No. NBP315486100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HMGB1/HMG-1 Polyclonal antibody specifically detects HMGB1/HMG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB1/HMG-1
Applications Western Blot, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Knockout Validated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Host Species Rabbit
Immunogen A synthetic acetylated peptide around K29 of human HMGB1.
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3146
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.