missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HMGB1 (HMG-1) Human ProcartaPlex™ Simplex Kit

Product Code. 17410723
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 17410723

Brand: Invitrogen™ EPX01012422901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Human HMGB1 (HMG-1) Simplex ProcartaPlex Kit measures HMGB1 (HMG-1) protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.

The Human HMGB1 (HMG-1) Simplex ProcartaPlex Kit measures HMGB1 (HMG-1) protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (e.g., when you are not using a pre-configured multiplex panel), only one buffer kit (Cat. No. EPX010-10420-901) is needed for each assay plate regardless of plex size.

Note that Human HMGB1 (HMG-1) Simplex ProcartaPlex Kit is not suitable for use with supernatant sample types.

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum and just four hours to obtain analyzed results.

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—the most referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample—from plasma, serum, and other bodily fluids.

The Luminex MagPlex superparamagnetic microsphere beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create 100 spectrally unique signatures that can be identified by the Luminex xMAP detection systems (Luminex 200, FLEXMAP 3D, and MAGPIX systems). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a magnetic bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Region 51
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
Format Simplex Kit
For Use With (Equipment) Luminex™ Instruments
Gene High mobility group box 1
Gene Alias HMG1, HMG3, HMG-1, SBP-1
Gene ID (Entrez) 3146
Gene Symbol HMGB1
Interassay CV Less than 15%
Intraassay CV Less than 15%
Product Line ProcartaPlex
Protein High mobility group protein B1
Sample Type Serum, Plasma
Sample Volume Serum, Plasma: 25 μL
UniProt ID P09429
Content And Storage

• 1 vial Capture Beads (50X)
• 1 vial Biotinylated Detection Antibody (50X)
• 2 vials Human Standard Mix 10 (lyophilized)

Store at 2°C to 8°C.

Research Area Oncology
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.