missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1/HMG-1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2322PE
This item is not returnable.
View return policy
Description
HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep samples. It is validated for Flow Cytometry, Flow (Intracellular), Knockdown Validated.
Specifications
| HMGB1/HMG-1 | |
| Polyclonal | |
| Flow Cytometry, Flow (Intracellular), Knockdown Validated | |
| P09429 | |
| HMGB1 | |
| A synthetic peptide made to an internal portion of the human HMGB1 protein sequence (between residues 100-200). [UniProt #P09429] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, rat, dog, bovine, sheep. Predicted to react with porcine, equine and rabbit based on 100% sequence homology. | |
| Store at 4C in the dark. |
| Flow Cytometry, KnockDown | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| Rabbit | |
| 29 kDa | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
| 3146 | |
| Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction