missing translation for 'onlineSavingsMsg'
Learn More

HMGB1/HMG-1 Antibody Pair, Novus Biologicals™

Product Code. 18391728 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391728 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391728 Supplier Novus Biologicals Supplier No. H00003146AP44

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pair

HMGB1/HMG-1 Monoclonal antibody specifically detects HMGB1/HMG-1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB1/HMG-1
Applications Sandwich ELISA
Classification Monoclonal
Conjugate Unconjugated
Dilution Sandwich ELISA 3 to 100 ng/mL
Formulation 1x PBS, pH 7.4
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Quantity 1 Set
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3146
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.