missing translation for 'onlineSavingsMsg'
Learn More

HMGB1/HMG-1 Antibody (951420) [DyLight 680], Novus Biologicals™

Product Code. 30763666 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763666 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763666 Supplier Novus Biologicals Supplier No. FAB16901Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

HMGB1/HMG-1 Monoclonal antibody specifically detects HMGB1/HMG-1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB1/HMG-1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 951420
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Host Species Rat
Immunogen Human HMGB1/HMG-1 synthetic peptide between amino acids Lys50-Ser100, Accession # P09429
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3146
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.