missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HMGB1/HMG-1 Monoclonal antibody specifically detects HMGB1/HMG-1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | HMGB1/HMG-1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 951420 |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein |
| Host Species | Rat |
| Immunogen | Human HMGB1/HMG-1 synthetic peptide between amino acids Lys50-Ser100, Accession # P09429 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?