missing translation for 'onlineSavingsMsg'
Learn More

HMGB1/HMG-1 Antibody (19N12A1), PE, Novus Biologicals™

Product Code. 18425881 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425881 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425881 Supplier Novus Biologicals Supplier No. NBP227400PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMGB1/HMG-1 Monoclonal antibody specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB1/HMG-1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 19N12A1
Conjugate PE
Formulation PBS
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Host Species Mouse
Immunogen Full-length recombinant human HMGB1 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3146
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.