missing translation for 'onlineSavingsMsg'
Learn More

HMGB1/HMG-1 Antibody (19N12A1), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18407096 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407096 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407096 Supplier Novus Biologicals Supplier No. NBP227400MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMGB1/HMG-1 Monoclonal antibody specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB1/HMG-1
Applications Flow Cytometry
Classification Monoclonal
Clone 19N12A1
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Host Species Mouse
Immunogen Full-length recombinant human HMGB1 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3146
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.