missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1/HMG-1 Antibody (1002147) [mFluor Violet 450 SE], Novus Biologicals™
Shop All Bio Techne ProductsDescription
HMGB1/HMG-1 Monoclonal antibody specifically detects HMGB1/HMG-1 in Human,Mouse samples. It is validated for Immunocytochemistry
Specifications
Specifications
| Antigen | HMGB1/HMG-1 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1002147 |
| Conjugate | mFluor Violet 450 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein |
| Host Species | Mouse |
| Immunogen | Synthetic peptide containing human HMGB1, Thr85-Gly166, Accession # P09429 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?