Learn More
HMGA2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20795-1-AP-150UL
This item is not returnable.
View return policy
Description
The HMGA2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human HMGA2. This antibody recognizes human, mouse antigen. The HMGA2 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Specifications
| HMGA2 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 8091 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP) | |
| 507 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido