missing translation for 'onlineSavingsMsg'
Learn More

HMGA1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18398724 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398724 100 μL 100µL
18042954 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18398724 Supplier Novus Biologicals Supplier No. NBP303907100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HMGA1 Polyclonal specifically detects HMGA1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGA1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias high mobility group AT-hook 1, High mobility group AT-hook protein 1, High mobility group protein A1, high mobility group protein HMG-I/HMG-Y, High mobility group protein R, high-mobility group (nonhistone chromosomal) protein isoforms I and Y, HMGA1A, HMG-I(Y), HMGIYMGC12816, HMG-R, MGC4242, MGC4854, nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HMGA1 (NP_665908.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3159
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.