missing translation for 'onlineSavingsMsg'
Learn More

HMGA1 Antibody (2A1), Novus Biologicals™

Product Code. 18390668 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390668 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390668 Supplier Novus Biologicals Supplier No. H00003159M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMGA1 Monoclonal antibody specifically detects HMGA1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGA1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2A1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_665906
Gene Alias high mobility group AT-hook 1, High mobility group AT-hook protein 1, High mobility group protein A1, high mobility group protein HMG-I/HMG-Y, High mobility group protein R, high-mobility group (nonhistone chromosomal) protein isoforms I and Y, HMGA1A, HMG-I(Y), HMGIYMGC12816, HMG-R, MGC4242, MGC4854, nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y
Host Species Mouse
Immunogen HMGA1 (NP_665906.1, 1 a.a. ~ 107 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSESSSKSSQPLASKQEKDGTEKRGRGRPRKQPPVSPGTALVGSQKEPSEVPTPKRPRGRPKGSKNKGAAKTRKTTTTPGRKPRGRPKKLEKEEEEGISQESSEEEQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3159
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.