missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12085-2-AP-20UL
This item is not returnable.
View return policy
Description
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.Specifications
| HMG20A | |
| Polyclonal | |
| Unconjugated | |
| HMG20A | |
| high mobility group 20A, HMG box containing protein 20A, HMG20A, HMGX1, HMGXB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10363, 315689, 66867 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9DC33, Q9NP66 | |
| Hmg20a | |
| HMG20A Fusion Protein Ag2718 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido