missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20A Antibody (OTI2H4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71596APC
This item is not returnable.
View return policy
Description
HMG20A Monoclonal antibody specifically detects HMG20A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| HMG20A | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2H4 | |
| Western Blot, Immunohistochemistry | |
| FLJ10739, high-mobility group 20A, HMG box domain containing 1, HMG box-containing protein 20A, HMG domain-containing protein 1, HMGX1HMG domain-containing protein HMGX1, HMGXB1high mobility group protein 20A | |
| Human recombinant protein fragment corresponding to amino acids 1-281 of human HMG20A(NP_060670) produced in E.coli. | |
| 0.1 mL | |
| Chromatin Research, Stem Cell Markers | |
| 10363 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction