missing translation for 'onlineSavingsMsg'
Learn More

HMG20A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen HMG20A
Dilution Western Blot 1:100 - 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30229591
View Documents
Novus Biologicals
NBP3-38406-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30230011
View Documents
Novus Biologicals
NBP3-38406-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HMG20A Polyclonal antibody specifically detects HMG20A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications

Specifications

HMG20A
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Chromatin Research, Stem Cell Markers
PBS (pH 7.3), 50% glycerol
10363
IgG
Affinity purified
Western Blot 1:100 - 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100
Polyclonal
Purified
RUO
Human, Mouse, Rat
FLJ10739, high-mobility group 20A, HMG box domain containing 1, HMG box-containing protein 20A, HMG domain-containing protein 1, HMGX1HMG domain-containing protein HMGX1, HMGXB1high mobility group protein 20A
Recombinant fusion protein containing a sequence corresponding to amino acids 1-347 of human HMG20A (NP_060670.1).,, Sequence:, MENLMTSSTLPPLFADEDGSKESNDLATTGLNHPEVPYSSGATSSTNNPEFVEDLSQGQLLQSESSNAAEGNEQRHEDEQRSKRGGWSKGRKRKKPLRDSNAPKSPLTGYVRFMNERREQLRAKRPEVPFPEITRMLGNEWSKLPPEEKQRYLDEADRDKERYMKELEQYQKTEAYKVFSRKTQDRQKGKSHRQDAARQATHDHEKETEVKERSVFDIPIFTEEFLNHSKAREAELRQLRKSNMEFEERNAALQKHVESMRTAVEKLEVDVIQERSRNTVLQQHLETLRQVLTSSFASMPLPGSGETPTVDTIDSYMNRLHSIILANPQDNENFIATVREVVNRLDR
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.