missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG-CoA Reductase/HMGCR Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP3-05292
This item is not returnable.
View return policy
Description
HMG-CoA Reductase/HMGCR Polyclonal antibody specifically detects HMG-CoA Reductase/HMGCR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Peptide ELISA
Specifications
| HMG-CoA Reductase/HMGCR | |
| Polyclonal | |
| Flow Cytometry 10 μg/mL, Immunocytochemistry/ Immunofluorescence 10 μg/mL, Peptide ELISA Detection limit 1:8000 | |
| 3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3 | |
| Peptide with sequence C-ADPSPQNSTADTSK corresponding to internal region according to NP_000850.1. | |
| 0.1 mg | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 3156 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunofluorescence, Peptide ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction