missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG-CoA Reductase/HMGCR Antibody (CL0259), Novus Biologicals™
Mouse Monoclonal Antibody
£292.00 - £451.00
Specifications
| Antigen | HMG-CoA Reductase/HMGCR |
|---|---|
| Clone | CL0259 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18272905
|
Novus Biologicals
NBP2-61616 |
100 μL |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658957
|
Novus Biologicals
NBP2-61616-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HMG-CoA Reductase/HMGCR Monoclonal specifically detects HMG-CoA Reductase/HMGCR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| HMG-CoA Reductase/HMGCR | |
| Monoclonal | |
| Purified | |
| Human | |
| 3156 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: HIPAYKLETLMETHERGVSIRRQLLSKKLSEPSSLQYLPYRDYNYSLVMGACCENVIGYMPIPVGVAGPLCLDEKEFQVPMATTEGCLVA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL0259 | |
| Unconjugated | |
| Mouse | |
| 3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3 | |
| HMGCR | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title