missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG-CoA Reductase/HMGCR Antibody (CL0259) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61616H
This item is not returnable.
View return policy
Description
HMG-CoA Reductase/HMGCR Monoclonal antibody specifically detects HMG-CoA Reductase/HMGCR in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| HMG-CoA Reductase/HMGCR | |
| Monoclonal | |
| HRP | |
| 3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3 | |
| This antibody was developed using a recombinant protein derived from P04035, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 3156 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL0259 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction