Learn More
HMCES Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560876
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: WRLLTMAGIF DCWEPPEGGD VLYSYTIITV DSCKGLSDIH HRMPAILDGE EAVSKWLDFG EVSTQEALKL IHPTENITFH AVSSVVNNS Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
HMCES Polyclonal antibody specifically detects HMCES in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
HMCES | |
Polyclonal | |
Unconjugated | |
HMCES | |
5-hydroxymethylcytosine (hmC) binding, ES cell specific; 5-hydroxymethylcytosine (hmC) binding, ES cell-specific; 5-hydroxymethylcytosine binding, ES cell specific; 8430410A17Rik; Abasic site processing protein HMCES; abasic site processing protein HMCES; embryonic stem cell-specific 5-hydroxymethylcytosine-binding protein; C20H3orf37; C22H3orf37; C3orf37; C85376; DC12; Embryonic stem cell-specific 5-hydroxymethylcytosine-binding protein; ES cell-specific 5hmC-binding protein; HMCES; Peptidase HMCES; Putative endonuclease HMCES; putative peptidase SRAPD1; SRAP domain-containing protein 1; Srap1; Srapd1; UPF0361 protein C3orf37; UPF0361 protein C3orf37 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56941 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96FZ2 | |
HMCES | |
Recombinant protein corresponding to Human HMCES | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only