missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMCES Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570309
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 79%; Rabbit: 100%; Rat: 93%.
Sensor of abasic sites in single-stranded DNA (ssDNA) required to preserve genome integrity by promoting error-free repair of abasic sites (PubMed:30554877, PubMed:31235915, PubMed:31235913). Acts as an enzyme that recognizes and binds abasic sites in ssDNA at replication forks and chemically modifies the lesion by forming a covalent cross-link with DNA: forms a stable thiazolidine linkage between a ring-opened abasic site and the alpha-amino and sulfhydryl substituents of its N-terminal catalytic cysteine residue (PubMed:30554877, PubMed:31235913). The HMCES DNA-protein cross-link is then degraded by the proteasome (PubMed:30554877). Promotes error-free repair of abasic sites by acting as a 'suicide' enzyme that is degraded, thereby protecting abasic sites from translesion synthesis (TLS) polymerases and endonucleases that are error-prone and would generate mutations and double-strand breaks (PubMed:30554877). Has preference for ssDNA, but can also accommodate double-stranded DNA with 3' or 5' overhang (dsDNA), and dsDNA-ssDNA 3' junction (PubMed:31235915, PubMed:31806351). Also involved in class switch recombination (CSR) in B-cells independently of the formation of a DNA-protein cross-link: acts by binding and protecting ssDNA overhangs to promote DNA double-strand break repair through the microhomology-mediated alternative-end-joining (Alt-EJ) pathway. Acts as a protease: mediates autocatalytic processing of its N-terminal methionine in order to expose the catalytic cysteine. [UniProt].
Specifications
| HMCES | |
| Polyclonal | |
| Unconjugated | |
| HMCES | |
| 5-hydroxymethylcytosine (hmC) binding, ES cell specific; 5-hydroxymethylcytosine (hmC) binding, ES cell-specific; 5-hydroxymethylcytosine binding, ES cell specific; 8430410A17Rik; Abasic site processing protein HMCES; abasic site processing protein HMCES; embryonic stem cell-specific 5-hydroxymethylcytosine-binding protein; C20H3orf37; C22H3orf37; C3orf37; C85376; DC12; Embryonic stem cell-specific 5-hydroxymethylcytosine-binding protein; ES cell-specific 5hmC-binding protein; HMCES; Peptidase HMCES; Putative endonuclease HMCES; putative peptidase SRAPD1; SRAP domain-containing protein 1; Srap1; Srapd1; UPF0361 protein C3orf37; UPF0361 protein C3orf37 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 232210 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8R1M0 | |
| HMCES | |
| synthetic peptide directed towards the middle region of human HMCES (aa 41-90). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction