missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMCES Antibody, Novus Biologicals™
Description
HMCES Polyclonal antibody specifically detects HMCES in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antigen | HMCES |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 - 1:10000, Immunoprecipitation 20-40 ug/mg lysate |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | chromosome 3 open reading frame 37, DC12, MGC111075 |
| Host Species | Rabbit |
| Immunogen | Recognizes region between residues Between between 66.. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?