Learn More
Invitrogen™ HMBS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595924
Description
Immunogen sequence: MSGNGNAAAT AEENSPKMRV IRVGTRKSQL ARIQTDSVVA TLKASYPGLQ FEIIAMSTTG DKILDTALSK IGEKSLFTKE LEHALEKNEV DLVVHSLKDL PTVLPPGFTI GAICKRENPH DAVVFHPKFV GKTLETLPEK SVVGTSSLRR AAQLQRKFPH LEFRSIRGNL NTRLRKLDEQ QEFSAIILAT AGLQRMGWHN RVGQILHPEE CMYAVGQGAL GVEVRAKDQD ILDLVGVLHD PETLLRCIAE RAFLRHLEGG CSVPVAVHTA MKDGQLYLTG GVWSLDGSDS IQETMQATIH VPAQHEDGPE DDPQLVGITA RNIPRGPQLA AQNLGISLAN LLLSKGAKNI LDVARQLNDA H; Positive Samples: HeLa, Raji, Mouse spleen; Cellular Location: Cytoplasm.
PBGD (porphobilinogen deaminase), also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.
Specifications
| HMBS | |
| Polyclonal | |
| Unconjugated | |
| Hmbs | |
| alternative name: porphobilinogen deaminase; HEM3; hemC; HMBS; Hydroxymethylbilane synthase; PBG D; PBGD; PBG-D; PORC; Porphobilinogen deaminase; porphyria, acute; Chester type; pre-uroporphyrinogen synthase; similar to Porphobilinogen deaminase (Hydroxymethylbilane synthase) (HMBS) (Pre-uroporphyrinogen synthase) (PBG-D); T25658; Ups; uroporphyrinogen I synthase; uroporphyrinogen I synthetase; URO-S; Uros1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15288, 3145 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 6.93 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08397, P22907 | |
| Hmbs | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.