missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMBS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562230
This item is not returnable.
View return policy
Description
Immunogen sequence: EFSAIILATA GLQRMGWHNR VGQILHPEEC MYAVGQGALG VEVRAKDQDI LDLVGVLHDP ETLLRCIAER AFLRHLEGGC SVPVAVHTAM KDGQLYLTGG VWSLDGSDSI QETMQATIHV PAQHEDGPED DPQLV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
PBGD (porphobilinogen deaminase), also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.
Specifications
| HMBS | |
| Polyclonal | |
| Unconjugated | |
| Hmbs | |
| alternative name: porphobilinogen deaminase; HEM3; hemC; HMBS; Hydroxymethylbilane synthase; PBG D; PBGD; PBG-D; PORC; Porphobilinogen deaminase; porphyria, acute; Chester type; pre-uroporphyrinogen synthase; similar to Porphobilinogen deaminase (Hydroxymethylbilane synthase) (HMBS) (Pre-uroporphyrinogen synthase) (PBG-D); T25658; Ups; uroporphyrinogen I synthase; uroporphyrinogen I synthetase; URO-S; Uros1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3145 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P08397 | |
| Hmbs | |
| Recombinant protein corresponding to Human HMBS. Recombinant protein control fragment (Product #RP-89537). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction