missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMBOX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541634
This item is not returnable.
View return policy
Description
Peptide sequence: ETMSHYTDEP RFTIEQIDLL QRLRRTGMTK HEILHALETL DRLDQEHSDK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.
HMBOX1 is located at the boundary of 8p12.3 and 8p21.1. HMBOX1 proteins are highly conserved in human, mouse, rat, chicken and Xenopus laevis. Functional HMBOX1::EGFP (enhanced green fluorescent protein) fusion protein revealed that HMBOX1 accumulated more in cytoplasm than in nucleus. HMBOX1 is a transcription repressor. Human HMBOX1 is widely expressed in 18 tissues, and it is highly expressed in pancreas. Hmbox1 is widely expressed in mouse pancreas and the expression of this gene can also be detected in pallium, hippocampus and hypothalamus.
Specifications
| HMBOX1 | |
| Polyclonal | |
| Unconjugated | |
| HMBOX1 | |
| AI451877; AI604847; F830020C16Rik; HMBOX1; HNF1LA; homeobox containing 1; homeobox telomere-binding protein 1; Homeobox-containing protein 1; homeobox-containing protein PBHNF; HOT1; PBHNF; RGD1306787; TAH1; Telomere-associated homeobox-containing protein 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 219150, 79618 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6NT76, Q8BJA3 | |
| HMBOX1 | |
| Synthetic peptide directed towards the N-terminal of human HMBOX1 (aa 11-60). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction