missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HM74A/PUMA-G/GPR109A/NIACR1 Antibody (MM0329-6S24), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12358JF646
This item is not returnable.
View return policy
Description
HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| HM74A/PUMA-G/GPR109A/NIACR1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HCAR2 | |
| Human recombinant HM74A/PUMA-G/GPR109A/NIACR1 | |
| 0.1 mL | |
| 338442 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| MM0329-6S24 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| GPR109A, HCA2, hydroxycarboxylic acid receptor 2, NIACR1 | |
| Rat | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction