missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HM74A/PUMA-G/GPR109A/NIACR1 Antibody (245106), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2760APCCY7
This item is not returnable.
View return policy
Description
HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal antibody specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Flow Cytometry
Specifications
| HM74A/PUMA-G/GPR109A/NIACR1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 245106 | |
| Flow Cytometry | |
| GPR109A, HCA2, hydroxycarboxylic acid receptor 2, NIACR1 | |
| NS0 mouse myeloma cell line transfected with human HM74A/GPR109A, Met1-Pro363, Accession # Q8TDS4 | |
| 0.5 mL | |
| GPCR | |
| 338442 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction