Learn More
Epredia™ HM 525 NX Cryostat Blade Holders
Use Epredia™ HM 525 NX blade holders with HM 525 NX and other Epredia™ cryostats.
Brand: Epredia™ 705460
Code : 50
Additional Details : Weight : 1.35000kg
Description
Standard Knife Carrier:
- Allows for insertion of standard knives, with clearance angle adjustment
- Can be moved horizontally on precision dovetail guide with quick clamp system
- Includes anti-roll plate
Disposable Blade Carrier EC:
- Allows for insertion of high- and low-profile blades, with clearance angle adjustment
- Can be moved on precision dovetail guide with quick clamp plate
- Includes anti-roll plate
Disposable Blade Carrier EV:
- Use with vacutome system
- Allows for insertion of high- and low-profile disposable blades
- Includes a vacuum anti-roll hood for stretching and disposing of section waste
- Features a slot that is opened and closed by a vacuum-controlled flap
Magnetic Disposable Blade Carrier MV:
- Use with vacutome system
- Allows for insertion of disposable blades
- Includes a vacuum anti-roll hood for stretching and disposing of section waste
- Features a slot that is opened and closed by a vacuum-controlled flap
Specifications
Standard Knife Holder | |
Cryostat Blade Holder | |
Epredia HM 525 NX Cryostat and older Microm HM 550 |