missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ HM 340E Electronic Rotary Microtome
Designed for precision and stability which yields quality ribbons even for difficult to cut paraffin sections, the Epredia™ HM 340E Electronic Microtome is an ergonomically designed, electronic rotary microtome with stepping-motor advance technology for precision and stability to provide superior sectioning results.
Brand: Epredia™ 905190A
Description
- Fast, safe, and comfortable use
- Selectable retraction
- Specimen advance (specimen moves toward the knife)
- Zero-orientation head
- Guide-rail guidance
- Menu driven control panel can be used on either side of instrument
- Section thickness ranging from 0.5 to 100 μm
Ordering Information:
- Blade holder/Knife holder must be ordered separately unless purchased as a package.
Includes:
- 100 mL Epredia™ Para/Gard™ paraffin repellent
- Standard tools
- Brushed aluminum cover plate
- Dust cover
- Instruction manual
Specifications
HM 340E Rotary Microtome Package | |
Standard tools, Brushed aluminum cover plate, Dust cover, Instruction manual, universal cassette clamp (715020S) and disposable blade carrier E (705800) | |
20.5 in. | |
11 in. | |
16 in. | |
62 lb. |
Rotary Microtome | |
0.5 to 100μm | |
52 cm | |
28 cm | |
41 cm | |
28 kg |