missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLX1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04033AF750
This item is not returnable.
View return policy
Description
HLX1 Polyclonal antibody specifically detects HLX1 in Human samples. It is validated for Western Blot
Specifications
| HLX1 | |
| Polyclonal | |
| Western Blot | |
| H2.0 (Drosophila)-like homeo box 1, H2.0-like homeo box 1, H2.0-like homeo box-1, H2.0-like homeobox, H2.0-like homeobox 1, H2.0-like homeobox protein, HB24, HLX1H2.0-like homeobox 1 (Drosophila), Homeobox protein HB24, Homeobox protein HLX1 | |
| The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human H2.0-like Homeobox using the numbering given in entry NP_068777.1 (GeneID 3142). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 3142 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction