missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLTF Recombinant Rabbit Monoclonal Antibody (8T0I2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538133
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, 293T, HepG2, Mouse lung, Mouse heart, Rat lung Immunogen sequence: TEAGSPTIML LSLKAGGVGL NLSAASRVFL MDPAWNPAAE DQCFDRCHRL GQKQEVIITK FIVKDSVEEN MLKIQNKKRE LAAGAFGTKK PNADEMKQAK INEIRTLIDL.
This gene encodes a member of the SWI/SNF family. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein contains a RING finger DNA binding motif. Two transcript variants encoding the same protein have been found for this gene. However, use of an alternative translation start site produces an isoform that is truncated at the N-terminus compared to the full-length protein.
Specifications
| HLTF | |
| Recombinant Monoclonal | |
| 1.04 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14527, Q6PCN7 | |
| HLTF | |
| A synthetic peptide corresponding to a sequence within amino acids 900-1009 of human HLTF (Q14527). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8T0I2 | |
| Unconjugated | |
| HLTF | |
| AF010600; BC057116; DNA-binding protein/plasminogen activator inhibitor 1 regulator; DNA-binding protein/plasminogen activator inhibitor-1 regulator; helicase like transcription factor; helicase-like transcription factor; HIP116; HIP116A; HLTF; HLTF1; mem; P113; RING finger protein 80; RING-type E3 ubiquitin transferase HLTF; RNF80; Smarca3; Snf2l3; SNF2-like 3; Sucrose nonfermenting protein 2-like 3; sucrose nonfermenting-like 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 3; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3; TNF-response element-binding protein; ZBU1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20585, 295568, 6596 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction