missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLTF Rabbit anti-Human, Mouse, Rat, Clone: 8T0I2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00
Specifications
| Antigen | HLTF |
|---|---|
| Clone | 8T0I2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
HLTF Monoclonal antibody specifically detects HLTF in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| HLTF | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Chromatin Research, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6596 | |
| IgG | |
| Affinity purified |
| 8T0I2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DNA-binding protein/plasminogen activator inhibitor 1 regulator, EC 3.6.1, EC 3.6.4.-, EC 6.3.2.-, helicase-like transcription factor, HIP116ARING finger protein 80, HIP116matrix associated, actin dependent regulator of chromatin, RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3, subfamily a, member 3, Sucrose nonfermenting protein 2-like 3, ZBU1 | |
| A synthetic peptide corresponding to a sequence within amino acids 900-1009 of human HLTF (Q14527). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title